WHEATON™ Tall Form Dram Vial
Tubular Dram Vials
Additional Details:
Weight:
2.00000kg
Specification
Color | Clear |
---|---|
Diameter (Metric) Outer | 20.25 mm |
Material | Borosilicate Glass (Type 1, Class B) |
Pre-slit | No |
Sterility | Non-sterile |
Volume (Metric) | 7 mL |
Autoclavable | Autoclavable |
Closure Type | Screw Cap |
Height (Metric) | 42 mm |
Quantity | 264/Pk. |
Type | Vial |
Marking Spot | No |