Code:
NEW
3M™ Attest™ Biological Indicator
3M™ Attest™ Biological Indicator monitors the effectiveness of the steam sterilization process and provide results within 48 hours.
Additional Details:
Weight:
0.90000kg
Specification
Model | 1262 |
---|---|
Product Type | Biological Indicator |
Includes | Brown Cap |
Quantity | 100/Pk. |
Sterilization Method | Steam |