Learn More
Berkshire™ Ultra-Seal™ 3000 Wipers
Artikelnummer. p-209985 Alle ansehen Berkshire Produkte
Change view
Click to view available options
Abmessungen (L x B):
9 x 9 in. (22,86 x 22,86 cm)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Abmessungen (L x B)
18770269 9 x 9 in. (22,86 x 22,86 cm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18770269 Lieferant Berkshire™ Lieferanten-Nr. US3000.0909.8
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Edge
For ISO Class 3 (Class 1) applications
  • Knitted in tube form from the highest quality, 100% continuous-filament polyester yarn, leaving only two raw edges (which are then sealed)
  • No unsealed edges mean less potential for abrasion and contamination
  • Durable, two-ply wiper offers two to three times the absorbency of single-ply polyester
  • Laundered and packaged in an ISO Class 4 (Class 10) environment
Bestellinformation
Basis weight: 134g/m2 (17-988-421); 177g/m2 (17-988-422).
Spezifikation
Produkttyp Cleanroom Wipe
Länge (angloamerikanisch) 9 in.
Länge (metrisch) 22.86 cm
Breite (angloamerikanisch) 9 in.
Breite (metrisch) 22.86 cm
Material Polyester Knit
Flächengewicht 145
Zertifizierung/Konformität RoHS 2002/05, REACH
Abmessungen (L x B) 9 x 9 in. (22,86 x 22,86 cm)
Zur Verwendung mit (Anwendung) Chamber cleaning and cmP processing
Lage 2-ply