Corning™ Filter Adapter Caps and Solid Overcap
CORNING™ Filter Adapter Caps and Solid Overcap are constructed of polyethylene and sterilized by gamma irradiation.
Overview
Code: 67
Additional Details:
Additional Details: Weight: 0.01168kg
Disclaimers: Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
Product Code. 10031422
Quantity Price
1 £ 115.0 / Pack of 20
Estimated Shipment
04-11-2025
Add to basket
Description and Specification
Specification
Material Polyethylene
No. per Case 100/Cs
No. per Pack 5/Pk
  • Overcaps made from polyethylene
  • Filter adapter caps have peel-away covers for inserting barbed disc filters for sterile venting with 0.2 μm membrane
  • Sterlized by gamma irradiation
  • Solid red overcap is used in closed systems or applications where venting is not necessary
Documentation
Technical documents