Code:
NEW
Fisherbrand™ Maxi Pipet Tips
For use with large volume pipets
Additional Details:
Weight:
1.17876kg
Specification
Volume (Metric) | 1000 to 5000 μL |
---|---|
Filtered | Non-filtered |
Sterility | Non-sterile |
Color | Natural |
Autoclavable | Autoclavable |
Length (English) | 4.6 in. |
Length (Metric) | 117 mm |
Format | Bulk |
Tip Style | Macro |
For Use With (Equipment) | Gilson Pipetman |
Graduated | No |
Material | Polypropylene |