68.49 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
Product Type | Protective Coveralls |
---|---|
Size | Large |
Gender | Unisex |
Material | SMS Fabric |
Color | White |
Disposable | Single-use |
Waist Style | Elastic |
Cuff Style | Knit |
Closure Type | Front Zipper |
Seam Type | Serged |
Sterility | Non-sterile |
Certifications/Compliance | CE Category III, EN 1073-2 Particulate Radioactive Contamination, EN 1149-5 - Electrostatic Properties, EN 13034:2005+A1:2009 Type 6 Liquid Splash, EN ISO 13982-1:2004 Type 5 Hazardous Dusts |
Certifications/Compliance (Europe) | CE Category III, EN 1073-2, EN 1149-5, EN 13034:2005+A1:2009 Type 6, EN ISO 13982-1:2004 Type 5 |