20.29 GBP valid until 2025-12-31
Use promo code "24332" to get your promotional price.
Use promo code "24332" to get your promotional price.
Design | Interlocking |
---|---|
Material | PPO |
Type | Vial Holder |
Holds | 40 Cryogenic Tubes |
Product Line | Nunc™ |
Quantity | 1/Pk., 15/Cs. |
Tube Array | 4 x 10 |