Code:
13
Trajan™ BPX5 GC Capillary Columns
MS premium, low bleed phase designed for robustness and is suitable for a wide range of applications
Additional Details:
Weight:
0.50000kg
Specification
Diameter (Metric) Inner | 0.32 mm |
---|---|
Stationary Phase | SGE™ BPX5 |
For Use With (Application) | Phenols Analysis,Solvents Analysis,Phenols,Amines,Pesticide Analysis,Herbicides Analysis |
For Use With (Application) | Phenols Analysis,Solvents Analysis,Phenols,Amines,Pesticide Analysis,Herbicides Analysis |
For Use With (Application) | Phenols Analysis,Solvents Analysis,Phenols,Amines,Pesticide Analysis,Herbicides Analysis |
For Use With (Application) | Phenols Analysis,Solvents Analysis,Phenols,Amines,Pesticide Analysis,Herbicides Analysis |
For Use With (Application) | Phenols Analysis,Solvents Analysis,Phenols,Amines,Pesticide Analysis,Herbicides Analysis |
For Use With (Application) | Phenols Analysis,Solvents Analysis,Phenols,Amines,Pesticide Analysis,Herbicides Analysis |
Particle Size | 0.25 μm |
Temperature Range | -40°C to +360/370°C |
Film Thickness | 0.25 μm |
Length (Metric) | 15 m |
Polarity | Non polar |