Falcon™ Round-Bottom Polystyrene Test Tubes
Constructed of optically clear polystyrene for easy viewing of sample. Falcon™ Round-Bottom Polystyrene Tubes are widely used and referenced in laboratory protocols.
Overview
86.13 GBP valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Code: 66
Additional Details:
Additional Details: Weight: 2.88000kg
Product Code. 10186360
Quantity Price
1 £ 94.12 / Pack of 1000
In stock, ready to ship: 99
In stock, ready to ship: 100
Add to basket
  • Description and Specification
  • Alternative Products
  • Special Offers
Description and Specification
Specification
Color Clear
Description 1400 x G
Capacity (Metric) 5 mL
Length (Metric) 75 mm
Material Polystyrene
Disposable Single-use
Packaging Bulk, 1000/Cs.
Sterility Non-sterile
Graduated No
Diameter (Metric) 12 mm
Closure Type Not Included
Product Type Test Tube
  • Widely used and referenced in laboratory protocols
  • Sterile, nonpyrogenicity tested to less than 0.1 EU/mL (Cat. No. 352008 nonsterile)
The foundation for consistent research results
  • Provide reliable containment of laboratory fluid samples
  • Polystyrene tubes are best suited for procedures requiring high optical clarity
  • Medical-style packaging materials
  • 1,400 RCF rating (RCF claims refer to Relative Centrifugal Force measured in g-force for materials with a specific gravity of 1.0, used in an appropriate rotor with correct cushion and safety precautions. Tubes used with organic solvents at temperatures below 0°C may have lower RCF ratings.)
  • Tube dimensions and volumes are approximate