DWK Life Sciences Wheaton™ Glass Sample Bottles
Ideal for collecting, storing and exhibiting samples in the lab or field. Wheaton™ Snap Cap Sample Bottles are manufactured from Wheaton™ 800 soda lime clear glass that conforms to USP Type III requirements.
Overview
242.25 GBP valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Code: A0
Additional Details:
Additional Details: Weight: 2.44950kg
Product Code. 10545755
Quantity Price
1 £ 255.0 / Pack of 144
Call for latest availability
+44 (0)1509 555 500
Add to basket
  • Description and Specification
  • Accessories and Spare Parts
Description and Specification
Specification
Capacity 0.4 oz., 12 mL
Capacity (Metric) 12 mL
Capacity (English) 0.4 oz.
Color Clear
Diameter (English) 0.86 in.
Material Glass
Closure Color Natural
Height (Metric) 51 mm
Height (English) 2 in.
Closure Material LDPE
Closure Size 22 mm
Diameter (Metric) 22 mm
Closure Type Snap
  • Packaged in corrugated trays with partitions
  • Pre-attached low density polyethylene snap caps
  • Available in a range of capacities and cap sizes
Accessories and Spare Parts