Thermo Scientific™ RNase AWAY™ Surface Decontaminant
Eliminate RNase and DNA from laboratory surfaces. This product reduces dependency on carcinogenic DEPC treatments and saves time spent baking glassware.
Overview
137.34 GBP valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Code: NEW
Additional Details:
Additional Details: Weight: 3.79980kg
Product Code. 11952385
Quantity Price
1 £ 206.0 / 4 L
In stock, ready to ship: 18
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
  • Safety and Handling
  • Special Offers
Description and Specification
Specification
pH 11
Composition Sodium Hydroxide with Water
Container Type Bottle
Volume (English) 1.06 gal.
Volume (Metric) 4 L
  • Use on pipettes, gel boxes or RNA or DNA prep areas
  • Leaves no residue to interfere with gel polymerization or staining
  • Chemically stable and nonabrasive
  • Contains no strong acids
Alternative Products
Documentation Safety and Handling
[Warning]
[{code=EYE DAM. 2, sequence=1, description=Serious eye damage/eye irritation Category 2}, {code=SKIN IRRIT. 2, sequence=2, description=Skin corrosion/irritation Category 2}]
[{code=H315, sequence=1, description=Causes skin irritation.}, {code=H319, sequence=2, description=Causes serious eye irritation.}]
[{code=P260, sequence=51, description=Do not breathe dust/fume/gas/mist/vapours/spray.}, {code=P264, sequence=52, description=Wash hands thoroughly after handling.}, {code=P280, sequence=53, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P330+P331, sequence=54, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P303+P361+P353, sequence=55, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P304+P340, sequence=56, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=57, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=58, description=Immediately call a POISON CENTER/doctor/.}, {code=P363, sequence=59, description=Wash contaminated clothing before reuse.}, {code=P405, sequence=60, description=Store locked up.}, {code=P501b, sequence=61, description=Dispose of contents/container in accordance with local/regional/national/international regulations.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : sodium hydroxide}]
[{code=GHS07, sequence=1, description=EXCLAMATION MARK}]
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315- Causes skin irritation.
  • H319- Causes serious eye irritation.
Precautionary Statement
  • P260- Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264- Wash hands thoroughly after handling.
  • P280- Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310- Immediately call a POISON CENTER/doctor/.
  • P363- Wash contaminated clothing before reuse.
  • P405- Store locked up.
  • P501b- Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental Information
  • MIXTURE LIST- Contains : sodium hydroxide
Special Offers