Heathrow Scientific™ Autoclavable Bags
Clear, imprinted Autoclavable Bags with blue biohazard labeling specifically designed for safe disposal.
Additional Details:
Weight:
3.80000kg
Specification
Content And Storage | Room Temperature |
---|---|
Description | Clear Autoclavable Bags |
Autoclavable | Autoclavable (121°C) |
Capacity (English) | 6.8 gal. |
Capacity (Metric) | 25.75 L |
Color | Clear |
Disposable | Single-use |
For Use With (Application) | Hazardous Waste Disposal |
For Use With (Equipment) | Autoclave |
Material | Polypropylene |
Sterility | Non-sterile |
Thickness (Metric) | 2 mm |
No. per Case | 200/Cs. |
No. per Pack | 50 Pk. |
Outer Packaging | Box |
Thickness (English) | 80 mil |
Dimensions (L x W) | 415 x 600 mm |
Dimensions (L x W x H) | 16.3 x 23.6 x 0.08 in. |
Length (English) | 16.3 in. |
Length (Metric) | 41.5 cm |
Width (English) | 23.6 in. |
Width (Metric) | 60 cm |
Weight (English) | 0.04 lb. |
Weight (Metric) | 0.02 kg |
Product Type | Autoclavable Bags |