89.39 GBP valid until 2025-08-24
Use promo code "24132" to get your promotional price.
Use promo code "24132" to get your promotional price.
Diameter (Metric)Top | 140 mm |
---|---|
Capacity (Metric) | 300 mL |
Material | HDPE, Conductive |
Height (Metric) | 150 mm |
Autoclavable | Not Autoclavable |
Color | Black |
Includes | Removable Sieve, Earthing Cable |
Thread Size (Metric) | S65 |
Product Type | Safety Funnel w/ Hinged Lid, Type 1 |