48.18 GBP valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Use promo code "24123" to get your promotional price.
Includes | 1 Safety Bottle Carrier vessel and attachable handle |
---|---|
Product Type | Bottle Holder |
Description | Safety Bottle Carrier, 1.5 L, Green |
Array | 1 x 1 |
Autoclavable | Not Autoclavable |
Color | Green |
Holds | Bottles |
Material | Polypropylene |
Positions | 1 |
Shape | Cylindrical |
Shelves | None |
Stackable | No |
Sterility | Non-sterile |
For Use With (Application) | Transportation of bottled regeants |
For Use With (Equipment) | 1.5 L bottles |
Size | 1.5 L |
Temperature (Metric) Range | -20 to 60°C |
Length (Metric) | 11.4 cm |
Width (Metric) | 11.4 cm |
Height (Metric) | 16.5 cm (25.4 cm Height, Handle Extended) |
Diameter (Metric) Opening | 11.4 cm |
Diameter (Metric) Inner | 11.4 cm |
Weight (Metric) | .25 kg |