Code:
NEW
Honeywell Fluka Buffer solution pH 7.0 (20°C), Green colored, potassium dihydrogen phosphate / disodium hydrogen phosphate, with fungicide, Honeywell Fluka™
Green colored, potassium dihydrogen phosphate / disodium hydrogen phosphate, with fungicide
Additional Details:
CAS Number:
1336-21-6
Weight:
10.03000kg
Quantity | 10 L |
---|
Chemical Identifiers
CAS | 1336-21-6 |
---|---|
Molecular Formula | H5NO |
Molecular Weight (g/mol) | 35.05 |
InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
Synonym | |
PubChem CID | 14923 |
ChEBI | CHEBI:18219 |
IUPAC Name | azanium;hydroxide |
SMILES | N.O |
Specification
Chemical Name or Material | Buffer solution |
---|---|
CAS | 1336-21-6 |
pH | 7.0 |
Quantity | 10 L |
Molecular Formula | H5NO |
MDL Number | MFCD00066650 |
UN Number | NONH for all modes of transport |
InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
SMILES | N.O |
IUPAC Name | azanium;hydroxide |
Molecular Weight (g/mol) | 35.05 |
PubChem CID | 14923 |
ChEBI | CHEBI:18219 |